Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACE SIRNA Assays ›
          Gene Symbol:
          ACE
          Gene Name:
          angiotensin I converting enzyme
          Gene Aliases:
          ACE1, CD143, DCP, DCP1, ICH, MVCD3
          Chromosome Location:
          Chr.17: 63477061 - 63498380 on Build GRCh38
          UniGene ID:
          Hs.298469
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103987

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000789.3 NP_000780.1 17 2519
          NM_001178057.1 NP_001171528.1 6 814
          NM_152830.2 NP_690043.1 6 814
          XM_006721737.3 XP_006721800.2 6 927
          GenBank mRNA AB208971.1 - 17 2509
          AK301988.1 - 6 785
          BM908222.1 - 6 826
          J04144.1 - 17 2507
          JQ927553.1 - 17 2519
          M26657.1 - 6 785
          M29981.1 - 6 863
          X16295.1 - 6 791

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          blood vessel remodeling
          angiotensin maturation
          angiotensin catabolic process in blood
          regulation of renal output by angiotensin
          neutrophil mediated immunity
          antigen processing and presentation of peptide antigen via MHC class I
          regulation of systemic arterial blood pressure by renin-angiotensin
          spermatogenesis
          regulation of blood pressure
          regulation of smooth muscle cell migration
          regulation of vasoconstriction
          mononuclear cell proliferation
          regulation of vasodilation
          hormone catabolic process
          peptide catabolic process
          beta-amyloid metabolic process
          arachidonic acid secretion
          heart contraction
          regulation of angiotensin metabolic process
          hematopoietic stem cell differentiation
          positive regulation of protein tyrosine kinase activity
          cell proliferation in bone marrow
          positive regulation of peptidyl-tyrosine autophosphorylation
          regulation of hematopoietic stem cell proliferation
          negative regulation of gap junction assembly
          positive regulation of peptidyl-cysteine S-nitrosylation
          endopeptidase activity
          carboxypeptidase activity
          drug binding
          metallopeptidase activity
          exopeptidase activity
          tripeptidyl-peptidase activity
          peptidyl-dipeptidase activity
          zinc ion binding
          chloride ion binding
          mitogen-activated protein kinase kinase binding
          bradykinin receptor binding
          mitogen-activated protein kinase binding
          metallodipeptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0