Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADAM15 SIRNA Assays ›
          Gene Symbol:
          ADAM15
          Gene Name:
          ADAM metallopeptidase domain 15
          Gene Aliases:
          MDC15
          Chromosome Location:
          Chr.1: 155051272 - 155062776 on Build GRCh38
          UniGene ID:
          Hs.312098
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104053

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADAM15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001261464.1 NP_001248393.1 6 636
          NM_001261465.1 NP_001248394.1 6 606
          NM_001261466.1 NP_001248395.1 6 558
          NM_003815.4 NP_003806.3 6 606
          NM_207191.2 NP_997074.1 6 606
          NM_207194.2 NP_997077.1 6 606
          NM_207195.2 NP_997078.1 6 606
          NM_207196.2 NP_997079.1 6 606
          NM_207197.2 NP_997080.1 6 606
          NR_048577.1 6 606
          NR_048578.1 6 606
          NR_048579.1 6 606
          GenBank mRNA AB209157.1 - 5 2137
          AK000667.1 - 6 592
          AK295614.1 - 6 578
          AK296925.1 - 6 557
          AK297468.1 - 6 479
          AK308172.1 - 6 529
          AY518542.1 - 6 491
          AY560593.1 - 6 592
          AY560594.1 - 6 592
          AY560595.1 - 6 592
          AY560596.1 - 6 592
          AY560597.1 - 6 592
          AY560598.1 - 6 592
          AY560599.1 - 6 592
          AY560600.1 - 6 592
          AY560601.1 - 6 592
          AY576417.1 - 6 592
          BC014566.2 - 6 572 2958341
          BT009764.1 - 6 491
          U41767.1 - 6 498
          U46005.1 - 6 491

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of cell-matrix adhesion
          proteolysis
          apoptotic process
          cell-matrix adhesion
          integrin-mediated signaling pathway
          male gonad development
          extracellular matrix disassembly
          negative regulation of cell growth
          negative regulation of cell migration
          collagen catabolic process
          tissue regeneration
          innate immune response
          cardiac epithelial to mesenchymal transition
          protein kinase C signaling
          negative regulation of receptor binding
          metalloendopeptidase activity
          integrin binding
          protein binding
          metallopeptidase activity
          zinc ion binding
          SH3 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0