Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IDE SIRNA Assays ›
          Gene Symbol:
          IDE
          Gene Name:
          insulin degrading enzyme
          Gene Aliases:
          INSULYSIN
          Chromosome Location:
          Chr.10: 92451684 - 92574484 on Build GRCh38
          UniGene ID:
          Hs.500546
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104071

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IDE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001165946.1 NP_001159418.1 12 1440
          NM_001322793.1 NP_001309722.1 22 2856
          NM_001322794.1 NP_001309723.1 22 2739
          NM_001322795.1 NP_001309724.1 23 3038
          NM_001322796.1 NP_001309725.1 22 3020
          NM_001322797.1 NP_001309726.1 12 1440
          NM_004969.3 NP_004960.2 22 2856
          NR_136399.1 23 2980
          XM_017016187.1 XP_016871676.1 22 2779
          XM_017016188.1 XP_016871677.1 22 3373
          XM_017016189.1 XP_016871678.1 23 2772
          XM_017016190.1 XP_016871679.1 22 2899
          GenBank mRNA AB209204.1 - 13 1505
          AK093287.1 - 12 1412
          AK302578.1 - 12 1393
          AK312810.1 - 22 2781
          AK316407.1 - 12 1412
          BC096336.1 - 22 2820 40008464
          BC096337.1 - 22 2820 40008465
          BC096339.3 - 22 2820 40008468
          BX648462.1 - 22 3021
          M21188.1 - 22 2837

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013159, XM_008760362, XM_006231315, XM_006231314, XM_006231313, and others.

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          insulin receptor signaling pathway
          determination of adult lifespan
          bradykinin catabolic process
          ubiquitin homeostasis
          protein processing
          positive regulation of protein oligomerization
          hormone catabolic process
          negative regulation of proteolysis
          viral entry into host cell
          beta-amyloid metabolic process
          protein homooligomerization
          protein homotetramerization
          protein heterooligomerization
          proteolysis involved in cellular protein catabolic process
          insulin metabolic process
          insulin catabolic process
          beta-amyloid binding
          virus receptor activity
          glycoprotein binding
          metalloendopeptidase activity
          receptor binding
          protein binding
          ATP binding
          zinc ion binding
          ATPase activity
          beta-endorphin binding
          peptide binding
          protein homodimerization activity
          ubiquitin binding
          insulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0