Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1D SIRNA Assays ›
          Gene Symbol:
          CACNA1D
          Gene Name:
          calcium voltage-gated channel subunit alpha1 D
          Gene Aliases:
          CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
          Chromosome Location:
          Chr.3: 53495049 - 53813151 on Build GRCh38
          UniGene ID:
          Hs.476358
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104334

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNA1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000720.3 NP_000711.1 42 5120
          NM_001128839.2 NP_001122311.1 40 5015
          NM_001128840.2 NP_001122312.1 41 5060
          XM_005265448.3 XP_005265505.1 40 4897
          XM_011534094.2 XP_011532396.1 43 5212
          XM_011534096.2 XP_011532398.1 40 5023
          XM_011534097.2 XP_011532399.1 41 5679
          XM_011534099.2 XP_011532401.1 39 4662
          XM_011534100.2 XP_011532402.1 41 5107
          XM_017007137.1 XP_016862626.1 43 5212
          XM_017007138.1 XP_016862627.1 43 5209
          XM_017007139.1 XP_016862628.1 43 5212
          XM_017007140.1 XP_016862629.1 42 5152
          XM_017007141.1 XP_016862630.1 42 5152
          XM_017007142.1 XP_016862631.1 42 5128
          XM_017007143.1 XP_016862632.1 42 5128
          XM_017007144.1 XP_016862633.1 42 5128
          XM_017007145.1 XP_016862634.1 41 5083
          GenBank mRNA AB209171.1 - 36 4021
          AK294397.1 - 2 298
          EU363339.1 - 40 4897
          M76558.1 - 41 5452
          M83566.1 - 42 5120

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          sensory perception of sound
          regulation of insulin secretion
          positive regulation of calcium ion transport
          regulation of atrial cardiac muscle cell membrane repolarization
          cardiac conduction
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during SA node cell action potential
          regulation of heart rate by cardiac conduction
          regulation of potassium ion transmembrane transporter activity
          regulation of potassium ion transmembrane transport
          voltage-gated calcium channel activity
          high voltage-gated calcium channel activity
          ankyrin binding
          metal ion binding
          alpha-actinin binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved SA node cell action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-w4k6p:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0