Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN3 SIRNA Assays ›
          Gene Symbol:
          PTPN3
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 3
          Gene Aliases:
          PTP-H1, PTPH1
          Chromosome Location:
          Chr.9: 109375694 - 109538395 on Build GRCh38
          UniGene ID:
          Hs.436429
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104541

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145368.1 NP_001138840.1 6, 7 529
          NM_001145369.1 NP_001138841.1 1, 2 2578
          NM_001145370.1 NP_001138842.1 1, 2 2578
          NM_002829.3 NP_002820.3 6, 7 529
          XM_006717197.3 XP_006717260.1 6, 7 638
          XM_006717199.3 XP_006717262.1 6, 7 663
          XM_006717201.3 XP_006717264.1 5, 6 470
          XM_006717202.3 XP_006717265.1 3, 4 232
          XM_006717203.3 XP_006717266.1 3, 4 191
          XM_006717204.3 XP_006717267.1 6, 7 565
          XM_011518888.2 XP_011517190.1 6, 7 599
          XM_011518889.2 XP_011517191.1 6, 7 1269
          XM_017014955.1 XP_016870444.1 6, 7 621
          XM_017014956.1 XP_016870445.1 5, 6 477
          XM_017014957.1 XP_016870446.1 6, 7 565
          GenBank mRNA AK295875.1 - 5, 6 449
          AK304434.1 - 6, 7 529
          AK316066.1 - 6, 7 529
          BC126117.1 - 5, 6 439 8991833
          BC143848.1 - 5, 6 439 9052363
          BX648253.1 - 1, 2 2579
          BX648735.1 - 1, 2 1013
          DQ104439.1 - 5, 6 417
          M64572.1 - 5, 6 440

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_001059757, XM_008763739, XM_006238190, XM_006238191, XM_006238192, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011207, AK052928, BC115635, BC115634.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          peptidyl-tyrosine dephosphorylation
          negative regulation of mitotic cell cycle
          negative regulation of membrane protein ectodomain proteolysis
          liver regeneration
          regulation of membrane depolarization during action potential
          regulation of sodium ion transmembrane transporter activity
          phosphotyrosine binding
          protein tyrosine phosphatase activity
          protein binding
          cytoskeletal protein binding
          sodium channel regulator activity
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0