Search
Search

CLCA2| Gene Symbol: CLCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006536.5 | NP_006527.1 | 4, 5 | 754 | ||
| XM_011542448.2 | XP_011540750.1 | 4, 5 | 696 | |||
| GenBank mRNA | AB026833.1 | - | 4, 5 | 633 | ||
| AF043977.1 | - | 4, 5 | 700 | |||
| AF127980.1 | - | 4, 5 | 592 | |||
| AK290348.1 | - | 4, 5 | 659 | |||
| AK296719.1 | - | 4, 5 | 678 | |||
| AK307937.1 | - | 4, 5 | 657 | |||
| BC041096.1 | - | 4, 5 | 655 | 5752653 | ||