Search
Search

CACNA1H| Gene Symbol: CACNA1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005407.1 | NP_001005407.1 | 34 | 7624 | ||
| NM_021098.2 | NP_066921.2 | 35 | 7642 | |||
| XM_005255652.4 | XP_005255709.1 | 34 | 7470 | |||
| XM_006720963.3 | XP_006721026.1 | 34 | 7503 | |||
| XM_006720964.3 | XP_006721027.1 | 35 | 7455 | |||
| XM_006720965.3 | XP_006721028.1 | 34 | 7437 | |||
| XM_011522724.2 | XP_011521026.1 | 31 | 6952 | |||
| XM_017023819.1 | XP_016879308.1 | 34 | 7464 | |||
| GenBank mRNA | AF051946.3 | - | 35 | 7473 | ||
| AF070604.1 | - | 1 | 1282 | |||
| AF073931.1 | - | 35 | 7642 | |||
| AK074965.1 | - | 12 | 3025 | |||