Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD34 SIRNA Assays ›
          Gene Symbol:
          CD34
          Gene Name:
          CD34 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 207886538 - 207911338 on Build GRCh38
          UniGene ID:
          Hs.374990
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10495

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD34
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025109.1 NP_001020280.1 3, 4 782
          NM_001773.2 NP_001764.1 3, 4 782
          GenBank mRNA AF202879.1 - 1, 2 27
          AF523361.1 - 3, 4 595
          AF538601.1 - 2, 3 433
          AK291529.1 - 3, 4 535
          AK293030.1 - 3, 4 559
          AK300835.1 - 4, 5 599
          BC039146.1 - 3, 4 532 4746591
          M81104.1 - 3, 4 781
          S53910.1 - 3, 4 614
          S53911.1 - 3, 4 614

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endothelial cell proliferation
          glomerular filtration
          endothelium development
          cell-matrix adhesion
          signal transduction
          regulation of blood pressure
          cell proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          single organismal cell-cell adhesion
          hemopoiesis
          negative regulation of blood coagulation
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-10 production
          mesangial cell-matrix adhesion
          paracrine signaling
          positive regulation of odontogenesis
          negative regulation of nitric oxide biosynthetic process
          positive regulation of angiogenesis
          cell motility
          regulation of immune response
          leukocyte migration
          transdifferentiation
          vascular wound healing
          hematopoietic stem cell proliferation
          positive regulation of transforming growth factor beta production
          positive regulation of granulocyte colony-stimulating factor production
          extracellular exosome assembly
          glomerular endothelium development
          stem cell proliferation
          metanephric glomerular mesangial cell differentiation
          negative regulation of cellular response to heat
          negative regulation of cellular response to hypoxia
          negative regulation of interleukin-2 secretion
          positive regulation of glial cell-derived neurotrophic factor secretion
          negative regulation of neuron death
          positive regulation of vasculogenesis
          transcription factor binding
          carbohydrate binding
          sulfate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0