Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173077.2 | NP_775100.1 | 3 | 273 | ||
XM_017003372.1 | XP_016858861.1 | 4 | 6380 | |||
GenBank mRNA | AJ422118.1 | - | 3 | 227 | ||
BC112076.1 | - | 3 | 273 | 8327544 | ||
BC112078.1 | - | 3 | 273 | 8327546 |