Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LONP1 SIRNA Assays ›
          Gene Symbol:
          LONP1
          Gene Name:
          lon peptidase 1, mitochondrial
          Gene Aliases:
          CODASS, LON, LONP, LonHS, PIM1, PRSS15, hLON
          Chromosome Location:
          Chr.19: 5691834 - 5720452 on Build GRCh38
          UniGene ID:
          Hs.350265
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 105664

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LONP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276479.1 NP_001263408.1 3, 4 358
          NM_001276480.1 NP_001263409.1 2, 3 322
          NM_004793.3 NP_004784.2 2, 3 643
          NR_076392.1 3, 4 341
          XM_011528441.2 XP_011526743.1 2, 3 686
          GenBank mRNA AK056366.1 - 2, 3 322
          AK096433.1 - 3, 4 371
          AK096626.1 - 3, 4 358
          AK127867.1 - 3, 4 423
          AK298531.1 - 3, 4 322
          AK309720.1 - 3, 4 463
          AL096720.1 - 2, 3 189
          BC000235.1 - 2, 3 522 3350958
          BC109218.1 - 2, 3 233
          U02389.1 - 2, 3 619

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          response to hypoxia
          misfolded or incompletely synthesized protein catabolic process
          mitochondrion organization
          aging
          response to hormone
          response to aluminum ion
          mitochondrial DNA metabolic process
          cellular response to oxidative stress
          chaperone-mediated protein complex assembly
          protein homooligomerization
          proteolysis involved in cellular protein catabolic process
          oxidation-dependent protein catabolic process
          regulation of mitochondrial DNA replication
          single-stranded DNA binding
          single-stranded RNA binding
          ATP-dependent peptidase activity
          serine-type endopeptidase activity
          protein binding
          ATP binding
          ADP binding
          sequence-specific DNA binding
          G-quadruplex DNA binding
          DNA polymerase binding
          mitochondrial light strand promoter anti-sense binding
          mitochondrial heavy strand promoter anti-sense binding
          mitochondrial light strand promoter sense binding
          mitochondrial heavy strand promoter sense binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bv2mb:80/100.66.77.44:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0