Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KLK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015596.1 | NP_056411.1 | 3 | 301 | ||
GenBank mRNA | AB108824.1 | - | 3 | 258 | ||
AB451465.1 | - | 3 | 258 | |||
AL050220.1 | - | 3 | 306 | |||
AY923171.1 | - | 3 | 301 | |||
AY923172.1 | - | 4 | 577 | |||
AY923173.1 | - | 4 | 577 | |||
AY923174.1 | - | 3 | 493 | |||
AY923175.1 | - | 3 | 493 | |||
BC069334.1 | - | 3 | 302 | 7262205 | ||
BC069543.1 | - | 3 | 301 | 7262217 |