Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAM SIRNA Assays ›
Gene Symbol:
PAM
Gene Name:
peptidylglycine alpha-amidating monooxygenase
Gene Aliases:
PAL, PHM
Chromosome Location:
Chr.5: 102754783 - 103031105 on Build GRCh38
UniGene ID:
Hs.369430
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106323
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000919.3
NP_000910.2
8, 9
1015
NM_001177306.1
NP_001170777.1
8, 9
1015
NM_001319943.1
NP_001306872.1
9, 10
1581
NM_138766.2
NP_620121.1
8, 9
1015
NM_138821.2
NP_620176.1
8, 9
1015
NM_138822.2
NP_620177.1
8, 9
1015
NR_033440.1
7, 8
957
XM_011543419.2
XP_011541721.1
9, 10
1135
XM_017009495.1
XP_016864984.1
8, 9
1011
XM_017009496.1
XP_016864985.1
8, 9
1011
XM_017009497.1
XP_016864986.1
9, 10
1135
XM_017009498.1
XP_016864987.1
8, 9
1011
XM_017009499.1
XP_016864988.1
8, 9
1011
XM_017009500.1
XP_016864989.1
8, 9
1011
XM_017009501.1
XP_016864990.1
8, 9
1011
XM_017009502.1
XP_016864991.1
8, 9
1011
XM_017009503.1
XP_016864992.1
8, 9
1011
XM_017009504.1
XP_016864993.1
8, 9
1112
XM_017009505.1
XP_016864994.1
9, 10
1135
XM_017009506.1
XP_016864995.1
8, 9
1011
XM_017009507.1
XP_016864996.1
9, 10
1135
XM_017009508.1
XP_016864997.1
8, 9
1011
XM_017009509.1
XP_016864998.1
8, 9
1011
XM_017009510.1
XP_016864999.1
8, 9
1115
XM_017009511.1
XP_016865000.1
8, 9
1011
XM_017009512.1
XP_016865001.1
8, 9
1011
XM_017009513.1
XP_016865002.1
8, 9
1011
XM_017009514.1
XP_016865003.1
8, 9
1011
XM_017009515.1
XP_016865004.1
8, 9
1011
XM_017009516.1
XP_016865005.1
8, 9
1011
GenBank mRNA
AB095007.1
-
8, 9
642
AF010472.1
-
7, 8
558
AK290158.1
-
8, 9
753
AK296521.1
-
7, 8
723
BC018127.1
-
8, 9
1015
3887394
BT007419.1
-
8, 9
642
M37721.1
-
8, 9
830
S75037.1
-
8, 9
722
S75038.1
-
8, 9
722
Back To Top
More Information
Panther Classification:
Molecular Function -
oxygenase
Gene Ontology Categories:
Function(s)
Process(es)
peptide amidation
response to hypoxia
long-chain fatty acid metabolic process
regulation of transcription from RNA polymerase II promoter
central nervous system development
heart development
lactation
response to pH
toxin metabolic process
protein amidation
ovulation cycle process
response to estradiol
regulation of actin cytoskeleton organization
odontogenesis
response to drug
response to copper ion
regulation of protein secretion
protein homooligomerization
response to glucocorticoid
oxidation-reduction process
maternal process involved in female pregnancy
limb development
peptidylglycine monooxygenase activity
peptidylamidoglycolate lyase activity
copper ion binding
calcium ion binding
protein binding
zinc ion binding
protein kinase binding
L-ascorbic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0