Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTGS1 SIRNA Assays ›
          Gene Symbol:
          PTGS1
          Gene Name:
          prostaglandin-endoperoxide synthase 1
          Gene Aliases:
          COX1, COX3, PCOX1, PES-1, PGG/HS, PGHS-1, PGHS1, PHS1, PTGHS
          Chromosome Location:
          Chr.9: 122369906 - 122395703 on Build GRCh38
          UniGene ID:
          Hs.201978
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106344

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTGS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000962.3 NP_000953.2 11 1744
          NM_001271164.1 NP_001258093.1 10 1600
          NM_001271165.1 NP_001258094.1 10 1693
          NM_001271166.1 NP_001258095.1 11 1859
          NM_001271367.1 NP_001258296.1 11 1604
          NM_001271368.1 NP_001258297.1 12 2081
          NM_080591.2 NP_542158.1 11 1633
          XM_005252105.3 XP_005252162.1 12 1919
          XM_011518875.2 XP_011517177.1 12 2180
          XM_011518876.2 XP_011517178.1 12 5790
          XM_017014929.1 XP_016870418.1 14 2154
          GenBank mRNA AK290022.1 - 11 1697
          AK295221.1 - 12 2066
          AK304403.1 - 10 1693
          BC029840.1 - 11 1716 5174923
          CD014048.1 - 10 1566
          CD014049.1 - 11 1570
          DQ180741.1 - 10 1757
          DQ180742.1 - 10 1757
          M59979.1 - 10 1669
          S36219.1 - 10 1553
          S36271.1 - 10 1664
          S78220.1 - 10 1664

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          lipid metabolic process
          xenobiotic metabolic process
          inflammatory response
          response to oxidative stress
          aging
          learning
          memory
          regulation of blood pressure
          response to organonitrogen compound
          negative regulation of norepinephrine secretion
          sensory perception of pain
          cyclooxygenase pathway
          negative regulation of epinephrine secretion
          maintenance of blood-brain barrier
          regulation of cell proliferation
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          response to corticosterone
          oxidation-reduction process
          response to fatty acid
          cellular oxidant detoxification
          peroxidase activity
          prostaglandin-endoperoxide synthase activity
          lipid binding
          heme binding
          metal ion binding
          dioxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0