Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 5 | 630 | ||
XM_011531336.1 | XP_011529638.1 | 5 | 908 | |||
XM_017029507.1 | XP_016884996.1 | 4 | 647 | |||
GenBank mRNA | AB209849.1 | - | 5 | 585 | ||
AK313467.1 | - | 5 | 619 | |||
BC009960.2 | - | 5 | 617 | 4300487 | ||
BC015768.1 | - | 5 | 634 | 4868206 | ||
U81379.3 | - | 5 | 627 | |||
U81380.2 | - | 5 | 627 | |||
Y09328.1 | - | 5 | 620 | |||
Y10659.1 | - | 5 | 630 |