Search
Search

ABCA2| Gene Symbol: ABCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001606.4 | NP_001597.2 | 10, 11 | 1581 | ||
| NM_212533.2 | NP_997698.1 | 10, 11 | 1573 | |||
| XM_006716996.3 | XP_006717059.1 | 9, 10 | 1515 | |||
| GenBank mRNA | AF178941.1 | - | 9, 10 | 1483 | ||
| AF327657.1 | - | 9, 10 | 1439 | |||
| BC029282.1 | - | 9, 10 | 1539 | |||
| BC064542.1 | - | 10, 11 | 1469 | |||
| BC144588.1 | - | 9, 10 | 1563 | |||