Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITPR1 SIRNA Assays ›
          Gene Symbol:
          ITPR1
          Gene Name:
          inositol 1,4,5-trisphosphate receptor type 1
          Gene Aliases:
          ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
          Chromosome Location:
          Chr.3: 4493348 - 4847840 on Build GRCh38
          UniGene ID:
          Hs.567295
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106733

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITPR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099952.2 NP_001093422.2 15 1639
          NM_001168272.1 NP_001161744.1 14 1594
          NM_002222.5 NP_002213.5 14 1594
          XM_005265109.2 XP_005265166.1 15 1639
          XM_005265110.2 XP_005265167.1 15 1639
          XM_006713131.2 XP_006713194.1 15 1639
          XM_011533681.1 XP_011531983.1 15 1639
          XM_011533682.2 XP_011531984.1 15 1565
          XM_011533683.2 XP_011531985.1 15 1639
          XM_011533684.1 XP_011531986.1 15 1639
          XM_011533685.1 XP_011531987.1 15 1639
          XM_011533686.1 XP_011531988.1 15 1639
          XM_011533687.1 XP_011531989.1 14 1594
          XM_011533688.1 XP_011531990.1 14 1594
          XM_011533690.1 XP_011531992.1 15 1639
          XM_011533691.1 XP_011531993.1 15 1639
          XM_011533692.2 XP_011531994.1 15 1639
          XM_017006357.1 XP_016861846.1 15 1639
          XM_017006358.1 XP_016861847.1 15 1639
          GenBank mRNA AK293752.1 - 9 825
          D26070.1 - 14 1500
          L38019.2 - 15 1536
          U23850.1 - 12 1244

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010585, AK049015, AK137477, X15373.

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          calcium ion transport
          signal transduction
          post-embryonic development
          regulation of autophagy
          platelet activation
          endoplasmic reticulum calcium ion homeostasis
          epithelial fluid transport
          inositol phosphate-mediated signaling
          regulation of insulin secretion
          negative regulation of calcium-mediated signaling
          voluntary musculoskeletal movement
          release of sequestered calcium ion into cytosol
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          regulation of cardiac conduction
          inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          protein binding
          calcium ion transmembrane transporter activity
          calcium-release channel activity
          calcium channel inhibitor activity
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0