Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199987.1 | NP_001186916.1 | 3 | 733 | ||
NM_002493.4 | NP_002484.1 | 4 | 826 | |||
NM_182739.2 | NP_877416.1 | 3 | 781 | |||
GenBank mRNA | BG220924.1 | - | 2 | 437 |