Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SORL1 SIRNA Assays ›
          Gene Symbol:
          SORL1
          Gene Name:
          sortilin related receptor 1
          Gene Aliases:
          C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250
          Chromosome Location:
          Chr.11: 121452203 - 121633762 on Build GRCh38
          UniGene ID:
          Hs.368592
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106970
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 417.00
          Your Price
          Online offer:
          417.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SORL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003105.5 NP_003096.1 43, 44 6020
          GenBank mRNA AK294404.1 - 14, 15 2158
          AK295090.1 - 20, 21 2810
          AK295878.1 - 23, 24 2961
          AK302589.1 - 20, 21 2632
          AK316340.1 - 14, 15 1740
          BC137171.1 - 43, 44 5943 9021168
          Y08110.1 - 43, 44 5971

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00268342_m1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to Golgi
          protein targeting
          protein targeting to lysosome
          lipid transport
          post-Golgi vesicle-mediated transport
          receptor-mediated endocytosis
          signal transduction
          cholesterol metabolic process
          regulation of smooth muscle cell migration
          negative regulation of protein binding
          negative regulation of protein oligomerization
          negative regulation of MAP kinase activity
          protein retention in Golgi apparatus
          positive regulation of protein catabolic process
          negative regulation of neurogenesis
          protein maturation
          positive regulation of protein exit from endoplasmic reticulum
          negative regulation of neuron death
          negative regulation of beta-amyloid formation
          positive regulation of choline O-acetyltransferase activity
          negative regulation of tau-protein kinase activity
          positive regulation of ER to Golgi vesicle-mediated transport
          positive regulation of early endosome to recycling endosome transport
          negative regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of protein localization to early endosome
          negative regulation of neurofibrillary tangle assembly
          positive regulation of endocytic recycling
          beta-amyloid binding
          transmembrane signaling receptor activity
          protein binding
          low-density lipoprotein particle binding
          ADP-ribosylation factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0