Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107026
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
14
1724
NM_001243226.2
NP_001230155.2
15
1514
NM_001243227.1
NP_001230156.1
13
1558
NM_001243228.1
NP_001230157.1
14
1668
NM_001243230.1
NP_001230159.1
13
1197
NM_001243231.1
NP_001230160.1
12
1158
NM_001243232.1
NP_001230161.1
10
1180
NM_001243233.1
NP_001230162.1
10
1121
NM_001243234.1
NP_001230163.1
7
753
NM_001243235.1
NP_001230164.1
7
753
NM_001243236.1
NP_001230165.1
7
769
NM_001306207.1
NP_001293136.1
13
1349
NM_001306208.1
NP_001293137.1
10
1180
NM_003199.2
NP_003190.1
14
1724
XM_005266739.3
XP_005266796.2
13
1364
XM_005266741.3
XP_005266798.1
14
1324
XM_005266743.4
XP_005266800.1
13
1185
XM_005266744.3
XP_005266801.1
13
1305
XM_005266745.3
XP_005266802.1
13
1167
XM_005266747.3
XP_005266804.1
12
1161
XM_005266749.3
XP_005266806.1
10
952
XM_005266752.4
XP_005266809.1
10
1522
XM_005266754.3
XP_005266811.1
9
946
XM_005266755.4
XP_005266812.1
10
981
XM_005266761.3
XP_005266818.1
7
1262
XM_006722536.2
XP_006722599.1
14
1647
XM_006722537.2
XP_006722600.1
14
1280
XM_006722538.2
XP_006722601.1
13
1302
XM_011526158.2
XP_011524460.1
13
2278
XM_011526160.2
XP_011524462.1
9
1331
XM_017025934.1
XP_016881423.1
13
1361
XM_017025935.1
XP_016881424.1
13
1361
XM_017025936.1
XP_016881425.1
13
1364
XM_017025937.1
XP_016881426.1
13
1118
XM_017025938.1
XP_016881427.1
14
1277
XM_017025939.1
XP_016881428.1
14
1324
XM_017025940.1
XP_016881429.1
14
1647
XM_017025941.1
XP_016881430.1
14
1280
XM_017025942.1
XP_016881431.1
14
1324
XM_017025943.1
XP_016881432.1
14
1324
XM_017025944.1
XP_016881433.1
13
1582
XM_017025945.1
XP_016881434.1
13
1582
XM_017025946.1
XP_016881435.1
13
1599
XM_017025947.1
XP_016881436.1
13
1185
XM_017025948.1
XP_016881437.1
13
1582
XM_017025949.1
XP_016881438.1
13
1302
XM_017025950.1
XP_016881439.1
13
1579
XM_017025951.1
XP_016881440.1
12
1158
XM_017025952.1
XP_016881441.1
10
1172
XM_017025953.1
XP_016881442.1
10
952
XM_017025954.1
XP_016881443.1
10
1533
XM_017025955.1
XP_016881444.1
9
1267
XM_017025956.1
XP_016881445.1
9
946
XM_017025957.1
XP_016881446.1
7
1262
GenBank mRNA
AB209741.1
-
9
832
AK095041.1
-
10
1180
AK096862.1
-
12
1158
AK122765.1
-
14
1724
AK299169.1
-
13
1558
AK300038.1
-
7
699
AK300612.1
-
10
1121
AK300636.1
-
9
851
AK301144.1
-
13
1335
AK315074.1
-
15
1656
AK316165.1
-
9
895
AL518903.3
-
7
769
BC031056.1
-
15
1475
BC125084.1
-
13
1138
40117795
BC125085.1
-
13
1138
40117798
FR748210.1
-
13
1112
FR748211.1
-
13
1112
FR748212.1
-
11
932
FR748213.1
-
11
932
FR748214.1
-
12
1040
FR748215.1
-
10
860
FR748216.1
-
13
1103
FR748217.1
-
12
986
FR748218.1
-
8
722
FR748219.1
-
10
899
FR748220.1
-
7
632
FR748221.1
-
7
632
FR748222.1
-
7
632
FR748223.1
-
5
464
M74718.1
-
7
584
M74719.1
-
14
1311
X52079.1
-
11
977
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0