Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VLDLR SIRNA Assays ›
          Gene Symbol:
          VLDLR
          Gene Name:
          very low density lipoprotein receptor
          Gene Aliases:
          CAMRQ1, CARMQ1, CHRMQ1, VLDL-R, VLDLRCH
          Chromosome Location:
          Chr.9: 2621679 - 2656103 on Build GRCh38
          UniGene ID:
          Hs.370422
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107088

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VLDLR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018056.2 NP_001018066.1 9 1667
          NM_001322225.1 NP_001309154.1 8 1544
          NM_001322226.1 NP_001309155.1 8 1544
          NM_003383.4 NP_003374.3 9 1667
          GenBank mRNA AB208822.1 - 10 1423
          AK092381.1 - 7 934
          AK095744.1 - 5 747
          AK289527.1 - 1 50
          BC136562.1 - 9 1388 9020552
          BC142653.1 - 9 1329 40148037
          BC144245.1 - 9 1388 9052765
          D16493.1 - 9 1321
          D16494.1 - 9 1252
          L20470.1 - 9 1642
          L22431.1 - 9 1364
          S73849.1 - 9 1336

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_013703, NM_001161420, AK076172, L33417, BC013622, and others.

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          lipid transport
          receptor-mediated endocytosis
          signal transduction
          nervous system development
          heart development
          response to nutrient
          memory
          cholesterol metabolic process
          ventral spinal cord development
          cerebral cortex development
          cellular response to insulin stimulus
          glycoprotein transport
          very-low-density lipoprotein particle clearance
          reelin-mediated signaling pathway
          cellular response to glucose starvation
          response to drug
          positive regulation of protein kinase activity
          dendrite morphogenesis
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to hypoxia
          positive regulation of dendrite development
          glycoprotein binding
          low-density lipoprotein receptor activity
          calcium ion binding
          protein binding
          very-low-density lipoprotein particle receptor activity
          apolipoprotein binding
          very-low-density lipoprotein particle binding
          glycoprotein transporter activity
          reelin receptor activity
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0