Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282656.1 | NP_001269585.1 | 14 | 1760 | ||
NM_001282657.1 | NP_001269586.1 | 13 | 1670 | |||
NM_003442.5 | NP_003433.3 | 14 | 1763 | |||
XM_005253121.1 | XP_005253178.1 | 14 | 1711 | |||
XM_005253122.1 | XP_005253179.1 | 14 | 1736 | |||
XM_005253127.1 | XP_005253184.1 | 15 | 1789 | |||
XM_011520349.2 | XP_011518651.1 | 14 | 2482 | |||
XM_017018252.1 | XP_016873741.1 | 13 | 1647 | |||
XM_017018253.1 | XP_016873742.1 | 13 | 1644 | |||
XM_017018254.1 | XP_016873743.1 | 14 | 1748 | |||
XM_017018255.1 | XP_016873744.1 | 14 | 1838 | |||
XM_017018256.1 | XP_016873745.1 | 13 | 1650 | |||
GenBank mRNA | AF071771.1 | - | 11 | 1383 | ||
AK291133.1 | - | 14 | 1716 | |||
AK297214.1 | - | 13 | 1615 | |||
AK313330.1 | - | 14 | 1749 | |||
BC020219.1 | - | 14 | 1762 | 4902668 |