Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FDXR SIRNA Assays ›
          Gene Symbol:
          FDXR
          Gene Name:
          ferredoxin reductase
          Gene Aliases:
          ADXR
          Chromosome Location:
          Chr.17: 74862497 - 74873031 on Build GRCh38
          UniGene ID:
          Hs.69745
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107319

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FDXR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258012.3 NP_001244941.2 11 1437
          NM_001258013.3 NP_001244942.2 12 1401
          NM_001258014.3 NP_001244943.2 11 1284
          NM_001258015.2 NP_001244944.1 10 1188
          NM_001258016.3 NP_001244945.2 10 1253
          NM_004110.5 NP_004101.3 11 1326
          NM_024417.4 NP_077728.3 11 1308
          NR_047576.2 11 1408
          XM_006721772.1 XP_006721835.1 11 1256
          XM_011524527.1 XP_011522829.1 11 1274
          XM_011524528.1 XP_011522830.1 11 1363
          XM_011524529.1 XP_011522831.1 11 1366
          XM_011524531.2 XP_011522833.1 10 1259
          XM_011524532.2 XP_011522834.1 10 1266
          XM_011524533.2 XP_011522835.1 10 1234
          GenBank mRNA AK094120.1 - 11 1586
          AK098648.1 - 11 1245
          AK223530.1 - 11 1238
          AK293208.1 - 12 1346
          AK293210.1 - 11 1345
          AK295307.1 - 11 1437
          AK298908.1 - 10 1131
          AK301779.1 - 11 1221
          AK301977.1 - 10 1253
          BC002960.1 - 11 1247 3537008
          BC018911.2 - 11 1254
          BC034763.1 - 11 1909
          BC063493.1 - 11 1246 5404814
          BT006684.1 - 11 1221
          J03826.1 - 11 1259

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          steroid biosynthetic process
          C21-steroid hormone biosynthetic process
          cholesterol metabolic process
          sterol metabolic process
          oxidation-reduction process
          ferredoxin-NADP+ reductase activity
          protein binding
          NADPH-adrenodoxin reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0