Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFATC4 SIRNA Assays ›
Gene Symbol:
NFATC4
Gene Name:
nuclear factor of activated T-cells 4
Gene Aliases:
NF-AT3, NF-ATC4, NFAT3
Chromosome Location:
Chr.14: 24366911 - 24379604 on Build GRCh38
UniGene ID:
Hs.77810
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107453
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFATC4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136022.2
NP_001129494.1
7
2189
NM_001198965.1
NP_001185894.1
6
2177
NM_001198966.2
NP_001185895.1
6
1831
NM_001198967.2
NP_001185896.1
7
2189
NM_001288802.1
NP_001275731.1
6
1980
NM_001320043.1
NP_001306972.1
7
2189
NM_004554.4
NP_004545.2
6
2177
XM_011536797.2
XP_011535099.1
6
2012
XM_011536798.1
XP_011535100.1
6
2102
XM_011536799.2
XP_011535101.1
6
2014
GenBank mRNA
AK293185.1
-
6
2052
AK300450.1
-
5
2313
AK302271.1
-
6
1817
AK310433.1
-
6
1980
AK310445.1
-
7
2161
AK311078.1
-
5
1545
AK311425.1
-
6
1205
BC008857.2
-
5
1678
BC053855.1
-
6
1993
5588610
EU887632.1
-
7
2161
EU887633.1
-
7
2161
EU887634.1
-
7
2161
EU887635.1
-
7
1977
EU887636.1
-
7
1977
EU887637.1
-
7
1977
EU887638.1
-
7
2030
EU887639.1
-
7
2030
EU887640.1
-
7
2030
EU887641.1
-
6
1856
EU887642.1
-
6
1856
EU887643.1
-
6
1856
EU887644.1
-
6
1980
EU887645.1
-
6
1980
EU887646.1
-
6
1980
EU887647.1
-
5
2102
EU887648.1
-
5
2102
EU887649.1
-
5
2102
EU887650.1
-
3
853
EU887651.1
-
3
853
EU887652.1
-
3
853
EU887653.1
-
1
327
EU887654.1
-
1
327
EU887655.1
-
1
327
L41066.1
-
6
1997
Back To Top
More Information
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
transcription from RNA polymerase II promoter
inflammatory response
heart development
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of Wnt signaling pathway
negative regulation of protein binding
positive regulation of tumor necrosis factor production
calcineurin-NFAT signaling cascade
cellular response to UV
negative regulation of chromatin binding
cellular respiration
positive regulation of transcription from RNA polymerase II promoter
regulation of synaptic plasticity
negative regulation of dendrite morphogenesis
smooth muscle cell differentiation
muscle cell development
cellular response to lithium ion
negative regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of synapse maturation
positive regulation of apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription coactivator activity
protein binding
transcription factor binding
peroxisome proliferator activated receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0