Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLIT2 SIRNA Assays ›
          Gene Symbol:
          SLIT2
          Gene Name:
          slit guidance ligand 2
          Gene Aliases:
          SLIL3, Slit-2
          Chromosome Location:
          Chr.4: 20251905 - 20620561 on Build GRCh38
          UniGene ID:
          Hs.29802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107531

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLIT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289135.2 NP_001276064.1 15 3447
          NM_001289136.2 NP_001276065.1 15 3435
          NM_004787.3 NP_004778.1 16 3459
          XM_005248211.3 XP_005248268.1 16 3471
          XM_006713986.3 XP_006714049.1 16 3471
          XM_011513909.2 XP_011512211.1 16 1499
          XM_011513910.2 XP_011512212.2 4 2351
          XM_017008845.1 XP_016864334.1 16 1486
          GenBank mRNA AB017168.1 - 16 1752
          AF055585.1 - 15 2080
          AF133270.1 - 15 1536
          AK308444.1 - 4 420
          BC117190.1 - 16 1627 40125741
          BC143978.1 - 15 1603 9052496

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          negative regulation of protein phosphorylation
          cell migration involved in sprouting angiogenesis
          negative regulation of leukocyte chemotaxis
          axon guidance
          motor neuron axon guidance
          negative regulation of lamellipodium assembly
          negative regulation of endothelial cell migration
          negative regulation of smooth muscle cell migration
          spinal cord development
          chemorepulsion involved in embryonic olfactory bulb interneuron precursor migration
          chemorepulsion involved in postnatal olfactory bulb interneuron migration
          corticospinal neuron axon guidance through spinal cord
          negative regulation of cell growth
          negative regulation of cell migration
          negative regulation of actin filament polymerization
          retinal ganglion cell axon guidance
          response to nutrient levels
          cellular response to hormone stimulus
          negative regulation of GTPase activity
          Roundabout signaling pathway
          positive regulation of apoptotic process
          negative regulation of vascular permeability
          branching morphogenesis of an epithelial tube
          axon extension involved in axon guidance
          negative regulation of inflammatory response
          positive regulation of axonogenesis
          negative chemotaxis
          induction of negative chemotaxis
          negative regulation of small GTPase mediated signal transduction
          response to cortisol
          apoptotic process involved in luteolysis
          negative regulation of chemokine-mediated signaling pathway
          cellular response to heparin
          negative regulation of smooth muscle cell chemotaxis
          negative regulation of mononuclear cell migration
          negative regulation of neutrophil chemotaxis
          negative regulation of monocyte chemotaxis
          negative regulation of retinal ganglion cell axon guidance
          negative regulation of cellular response to growth factor stimulus
          GTPase inhibitor activity
          calcium ion binding
          protein binding
          heparin binding
          identical protein binding
          protein homodimerization activity
          laminin-1 binding
          proteoglycan binding
          heparan sulfate proteoglycan binding
          Roundabout binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ht48x:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0