Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107543
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
283.00
Your Price
Online offer:
274.65
274.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
24
7632
NM_001320156.1
NP_001307085.1
3
2143
NM_001320157.1
NP_001307086.1
3
2134
NM_004801.4
NP_004792.1
22
7422
NM_138735.2
NP_620072.1
6
3647
XM_005264642.3
XP_005264699.1
23
7524
XM_005264643.3
XP_005264700.1
22
7479
XM_006712137.3
XP_006712200.1
22
7434
XM_011533167.2
XP_011531469.1
23
7533
XM_011533171.2
XP_011531473.1
23
7512
XM_011533172.2
XP_011531474.1
22
7506
XM_011533174.2
XP_011531476.1
22
7488
XM_011533175.2
XP_011531477.1
21
7476
XM_011533177.2
XP_011531479.1
22
7443
XM_011533178.2
XP_011531480.1
20
7443
XM_011533183.1
XP_011531485.1
17
5215
XM_017005303.1
XP_016860792.1
24
7560
XM_017005304.1
XP_016860793.1
24
7557
XM_017005305.1
XP_016860794.1
24
7551
XM_017005306.1
XP_016860795.1
23
7548
XM_017005307.1
XP_016860796.1
23
7542
XM_017005308.1
XP_016860797.1
24
7539
XM_017005309.1
XP_016860798.1
23
7533
XM_017005310.1
XP_016860799.1
22
7530
XM_017005311.1
XP_016860800.1
23
7515
XM_017005312.1
XP_016860801.1
23
7509
XM_017005313.1
XP_016860802.1
23
7503
XM_017005314.1
XP_016860803.1
21
7500
XM_017005315.1
XP_016860804.1
22
7497
XM_017005316.1
XP_016860805.1
22
7497
XM_017005317.1
XP_016860806.1
22
7491
XM_017005318.1
XP_016860807.1
22
7488
XM_017005319.1
XP_016860808.1
22
7485
XM_017005320.1
XP_016860809.1
22
7476
XM_017005321.1
XP_016860810.1
23
7470
XM_017005322.1
XP_016860811.1
23
7461
XM_017005323.1
XP_016860812.1
19
7446
XM_017005324.1
XP_016860813.1
21
7416
XM_017005325.1
XP_016860814.1
21
7407
XM_017005326.1
XP_016860815.1
21
7404
XM_017005327.1
XP_016860816.1
21
7389
XM_017005328.1
XP_016860817.1
20
7371
XM_017005330.1
XP_016860819.1
18
7347
XM_017005331.1
XP_016860820.1
18
7332
XM_017005332.1
XP_016860821.1
17
7311
XM_017005333.1
XP_016860822.1
17
7302
XM_017005334.1
XP_016860823.1
20
5381
XM_017005335.1
XP_016860824.1
7
8727
XM_017005336.1
XP_016860825.1
7
8719
XM_017005337.1
XP_016860826.1
6
8628
GenBank mRNA
AB011150.2
-
21
6187
AB035356.1
-
21
6123
BC150247.1
-
21
6208
9007472
BX647616.1
-
3
2089
DA153976.1
-
1
539
DA168874.1
-
1
205
EF539882.1
-
24
7495
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline