Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KMT2A SIRNA Assays ›
          Gene Symbol:
          KMT2A
          Gene Name:
          lysine methyltransferase 2A
          Gene Aliases:
          ALL-1, CXXC7, HRX, HTRX1, MLL, MLL-AF9, MLL/GAS7, MLL1, MLL1A, TET1-MLL, TRX1, WDSTS
          Chromosome Location:
          Chr.11: 118436490 - 118526832 on Build GRCh38
          UniGene ID:
          Hs.258855
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107891

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KMT2A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001197104.1 NP_001184033.1 3 1513
          NM_005933.3 NP_005924.2 3 1513
          XM_011542829.2 XP_011541131.1 4 1604
          XM_011542830.2 XP_011541132.1 4 1604
          XM_011542831.2 XP_011541133.1 4 1604
          GenBank mRNA D14540.1 - 3 1513
          L04284.1 - 3 1490
          L04731.1 - 3 1304

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA methylation
          transcription from RNA polymerase II promoter
          protein complex assembly
          apoptotic process
          negative regulation of cell proliferation
          visual learning
          post-embryonic development
          anterior/posterior pattern specification
          peptidyl-lysine monomethylation
          positive regulation of transporter activity
          circadian regulation of gene expression
          embryonic hemopoiesis
          exploration behavior
          response to potassium ion
          histone H4-K16 acetylation
          histone H3-K4 dimethylation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of short-term neuronal synaptic plasticity
          spleen development
          homeostasis of number of cells within a tissue
          histone H3-K4 methylation
          positive regulation of histone H3-K4 methylation
          membrane depolarization
          definitive hemopoiesis
          regulation of histone H3-K14 acetylation
          histone H3-K4 trimethylation
          regulation of histone H3-K27 acetylation
          regulation of histone H3-K9 acetylation
          positive regulation of cellular response to drug
          core promoter sequence-specific DNA binding
          AT DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          zinc ion binding
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          identical protein binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          unmethylated CpG binding
          lysine-acetylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-29f27:80/100.66.78.233:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0