Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006593.3 | NP_006584.1 | 2, 3 | 1172 | ||
GenBank mRNA | AK226050.1 | - | 1, 2 | 1508 | ||
AK299533.1 | - | 1, 2 | 72 | |||
AK312567.1 | - | 2, 3 | 947 | |||
AK315865.1 | - | 1, 2 | 67 | |||
AK316161.1 | - | 1, 2 | 203 | |||
BC029289.1 | - | 2, 3 | 933 | |||
BC104844.1 | - | 2, 3 | 1100 | 8143847 | ||
BC113418.1 | - | 2, 3 | 1100 | 8322470 | ||
U49250.1 | - | 2, 3 | 879 |