Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HOXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006898.4 | NP_008829.3 | 3 | 1898 | ||
XM_005246509.3 | XP_005246566.1 | 4 | 2165 | |||
XM_005246510.4 | XP_005246567.1 | 4 | 1981 | |||
XM_005246511.3 | XP_005246568.1 | 4 | 2589 | |||
XM_005246513.4 | XP_005246570.1 | 4 | 2192 | |||
XM_006712477.2 | XP_006712540.1 | 3 | 1875 | |||
XM_011511065.2 | XP_011509367.1 | 4 | 3969 | |||
XM_011511066.2 | XP_011509368.1 | 4 | 2043 | |||
GenBank mRNA | BC005124.2 | - | 3 | 1899 | 3936607 | |
BC008789.2 | - | 2 | 1830 | 3628279 |