Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPE SIRNA Assays ›
          Gene Symbol:
          RPE RPEL1
          Gene Name:
          ribulose-5-phosphate-3-epimerase
          ribulose-5-phosphate-3-epimerase like 1
          Gene Aliases:
          RPE2-1
          -
          Chromosome Location:
          Chr.2: 210002565 - 210029156 on Build GRCh38
          Chromosome Location:
          Chr.10: 103245887 - 103248016 on Build GRCh38
          UniGene ID:
          Hs.282260
          Hs.458437
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 108134

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278282.1 NP_001265211.1 5, 6 587
          NM_001278283.1 NP_001265212.1 6, 7 667
          NM_001278286.1 NP_001265215.1 4, 5 511
          NM_001278288.1 NP_001265217.1 3, 4 502
          NM_001278289.1 NP_001265218.1 4, 5 582
          NM_001318926.1 NP_001305855.1 5, 6 636
          NM_001318927.1 NP_001305856.1 4, 5 582
          NM_001318929.1 NP_001305858.1 4, 5 582
          NM_001318930.1 NP_001305859.1 4, 5 571
          NM_001318931.1 NP_001305860.1 4, 5 533
          NM_006916.2 NP_008847.1 4, 5 556
          NM_199229.2 NP_954699.1 4, 5 582
          XM_006712677.3 XP_006712740.1 5, 6 529
          GenBank mRNA AJ224326.1 - 3, 4 324
          AK056028.1 - 4, 5 493
          AK093658.1 - 6, 7 598
          AK291035.1 - 4, 5 509
          AK308741.1 - 4, 5 420
          AL833120.1 - 4, 5 505
          BC005148.2 - 4, 5 490 3505010
          BC016764.1 - 4, 5 519 4107315
          BC072401.1 - 4, 5 491 6047438
          BG546879.1 - 4, 5 519
          BX403091.2 - 5, 6 542
          BX643719.1 - 4, 5 508
          CN372165.1 - 4, 5 505
          CR992340.1 - 4, 5 482
          DA237285.1 - 3, 4 429
          DA251418.1 - 1 493
          DA336224.1 - 4, 5 442
          Gene Symbol: RPEL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143909.1 NP_001137381.1 1 595
          GenBank mRNA BC137242.1 - 1 534 9021239
          BC137243.1 - 1 534 9021240
          BC157829.1 - 1 534 9057048
          BC157875.1 - 1 596 9057093
          BC171806.1 - 1 534 9054460
          BC171885.1 - 1 597 9054539

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008767287, XM_006245106, XM_008767289, XM_006245109, XM_006245105, and others.
          This siRNA also targets the following transcripts in Mouse: NM_025683, AK029088, AK150008, AK017774, BC006953, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          pentose-phosphate shunt
          pentose-phosphate shunt, non-oxidative branch
          pentose catabolic process
          cellular carbohydrate metabolic process
          ribulose-phosphate 3-epimerase activity
          identical protein binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rwjz2:80/100.66.72.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0