Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 598 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 405 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 412 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 301 | |||
NM_013269.5 | NP_037401.1 | 5 | 516 | |||
NR_036693.2 | 4 | 487 | ||||
GenBank mRNA | AF133299.1 | - | 5 | 502 | ||
AF285087.1 | - | 5 | 494 | |||
AF285088.1 | - | 3 | 307 | |||
AF285089.1 | - | 5 | 508 | |||
AK310150.1 | - | 7 | 1202 | |||
AK310554.1 | - | 4 | 478 | |||
AK313554.1 | - | 5 | 506 | |||
BC019883.1 | - | 5 | 1513 | 4524891 | ||
BU929933.1 | - | 5 | 501 | |||
DB124897.1 | - | 5 | 512 | |||
FN813349.1 | - | 4 | 385 | |||
FN813350.1 | - | 4 | 378 | |||
FN813351.1 | - | 3 | 274 | |||
KF958454.1 | - | 5 | 564 | |||
KF958455.1 | - | 6 | 566 | |||
KF958456.1 | - | 7 | 689 | |||
KF958457.1 | - | 5 | 564 | |||
KF958458.1 | - | 6 | 613 | |||
KF958459.1 | - | 3 | 349 | |||
KF971856.1 | - | 5 | 489 | |||
KF971857.1 | - | 6 | 571 | |||
KF971858.1 | - | 4 | 378 | |||
KF971859.1 | - | 4 | 385 |