Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NDOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144026.2 | NP_001137498.1 | 13 | 1700 | ||
NM_001144027.2 | NP_001137499.1 | 11, 12 | 1571 | |||
NM_001144028.2 | NP_001137500.1 | 12, 13 | 1652 | |||
NM_014434.3 | NP_055249.1 | 12, 13 | 1673 | |||
XM_011518545.1 | XP_011516847.1 | 12 | 1588 | |||
XM_011518547.1 | XP_011516849.1 | 13 | 1490 | |||
XM_011518548.2 | XP_011516850.1 | 12 | 2890 | |||
XM_017014626.1 | XP_016870115.1 | 13 | 1700 | |||
XM_017014627.1 | XP_016870116.1 | 12, 13 | 1673 | |||
GenBank mRNA | AF199509.1 | - | 12, 13 | 1609 | ||
AK026089.1 | - | 13 | 1672 | |||
AK074403.1 | - | 11, 12 | 1532 | |||
AK225995.1 | - | 13 | 1671 | |||
AK290026.1 | - | 12, 13 | 1670 | |||
AY077845.1 | - | 13 | 1618 | |||
BC093782.1 | - | 12, 13 | 1590 | 7939627 | ||
BC111943.1 | - | 12, 13 | 1590 | 8327411 |