Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108567
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
7
1242
NM_001256235.1
NP_001243164.1
8
1195
NM_001256236.1
NP_001243165.1
8
1437
NM_001256237.1
NP_001243166.1
7
1333
NM_001256238.1
NP_001243167.1
7
1241
NM_001256239.1
NP_001243168.1
6
1172
NM_001256240.1
NP_001243169.1
6
1184
NM_001283038.1
NP_001269967.1
7
1254
NM_001283039.1
NP_001269968.1
7
1207
NM_001283040.1
NP_001269969.1
5
826
NM_014489.3
NP_055304.1
7
1367
NR_027016.2
5
1009
NR_027017.2
8
1541
NR_027018.2
8
1206
NR_045923.1
6
1079
NR_045925.1
6
989
NR_045926.1
6
1001
NR_045927.1
6
1158
NR_045929.1
6
1125
NR_104270.1
6
1067
NR_104271.1
8
1327
NR_104272.1
5
997
XM_006718181.3
XP_006718244.1
6
1845
XM_006718183.3
XP_006718246.1
8
1262
XM_006718185.2
XP_006718248.1
7
1154
XM_006718186.2
XP_006718249.1
8
1314
XM_006718190.3
XP_006718253.1
7
1470
XM_006718191.3
XP_006718254.1
7
1458
XM_006718192.1
XP_006718255.1
9
1388
XM_006718193.2
XP_006718256.1
8
1195
XM_011519990.2
XP_011518292.2
8
1721
XM_011519991.2
XP_011518293.2
8
1712
XM_011519992.1
XP_011518294.1
9
1493
XM_011519993.2
XP_011518295.2
8
1819
XM_011519994.2
XP_011518296.2
8
1813
XM_011519995.2
XP_011518297.1
8
1644
XM_011519996.1
XP_011518298.1
9
1445
XM_011519997.1
XP_011518299.1
9
1786
XM_011519998.2
XP_011518300.1
9
1464
XM_011519999.1
XP_011518301.1
9
1423
XM_011520000.1
XP_011518302.1
8
1337
XM_011520001.1
XP_011518303.1
7
1129
XM_011520002.1
XP_011518304.1
7
1117
XM_011520003.2
XP_011518305.1
8
1653
XM_011520004.2
XP_011518306.1
8
1328
XM_011520005.2
XP_011518307.1
7
1225
XM_011520006.2
XP_011518308.1
8
1609
XM_011520007.2
XP_011518309.1
9
1378
XM_017017556.1
XP_016873045.1
7
1208
XM_017017557.1
XP_016873046.1
7
1277
XM_017017558.1
XP_016873047.1
7
1451
XM_017017559.1
XP_016873048.1
8
1314
XM_017017560.1
XP_016873049.1
8
1302
GenBank mRNA
AF159615.1
-
7
1369
AL049261.1
-
3
2398
AL096753.1
-
8
1192
BC009930.2
-
7
1130
3506614
BI858175.1
-
2
527
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz59j:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0