Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015423.2 | NP_056238.2 | 2, 3 | 563 | ||
GenBank mRNA | AF151057.1 | - | 2, 3 | 583 | ||
AF151838.1 | - | 2, 3 | 583 | |||
AF201943.1 | - | 2, 3 | 532 | |||
AF302110.1 | - | 2, 3 | 583 | |||
AK312529.1 | - | 2, 3 | 467 | |||
AL050073.1 | - | 1, 2 | 124 | |||
BC015470.1 | - | 2, 3 | 454 | 3914010 | ||
BC016728.1 | - | 2, 3 | 483 | 4081713 |