Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF2 SIRNA Assays ›
Gene Symbol:
IKZF2
Gene Name:
IKAROS family zinc finger 2
Gene Aliases:
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location:
Chr.2: 212999684 - 213152753 on Build GRCh38
UniGene ID:
Hs.604950
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108834
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079526.1
NP_001072994.1
9
1510
NM_016260.2
NP_057344.2
8
1441
XM_005246384.4
XP_005246441.1
8
1551
XM_005246385.3
XP_005246442.1
8
1581
XM_005246386.4
XP_005246443.1
8
1432
XM_011510802.2
XP_011509104.1
9
1623
XM_011510803.2
XP_011509105.1
9
1472
XM_011510804.2
XP_011509106.1
9
1395
XM_011510805.2
XP_011509107.1
9
1484
XM_011510807.2
XP_011509109.1
9
1659
XM_011510808.2
XP_011509110.1
9
1509
XM_011510809.2
XP_011509111.1
9
1521
XM_011510810.2
XP_011509112.1
9
1435
XM_011510811.2
XP_011509113.1
8
1556
XM_011510812.2
XP_011509114.1
8
1417
XM_011510814.2
XP_011509116.1
8
1915
XM_011510815.2
XP_011509117.1
8
1363
XM_011510816.2
XP_011509118.1
6
1364
XM_011510817.2
XP_011509119.1
7
1299
XM_011510818.2
XP_011509120.1
6
7018
XM_011510819.2
XP_011509121.1
7
1288
XM_017003589.1
XP_016859078.1
8
1503
XM_017003590.1
XP_016859079.1
8
1473
XM_017003591.1
XP_016859080.1
6
1906
XM_017003592.1
XP_016859081.1
7
1220
GenBank mRNA
AF130863.1
-
8
1441
AK301503.1
-
8
1437
AY008294.1
-
2
1905
AY587062.1
-
9
1425
AY587063.1
-
9
1387
AY587064.1
-
9
1471
AY587065.1
-
8
1278
AY587066.1
-
6
1050
AY587067.1
-
7
1089
AY587068.1
-
8
1349
AY587069.1
-
7
1194
AY587070.1
-
5
837
AY587071.1
-
6
1056
AY587072.1
-
3
384
AY587073.1
-
8
1451
BC028936.1
-
9
1510
4540576
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
biological_process
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0