Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF2 SIRNA Assays ›
Gene Symbol:
IKZF2
Gene Name:
IKAROS family zinc finger 2
Gene Aliases:
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location:
Chr.2: 212999684 - 213152753 on Build GRCh38
UniGene ID:
Hs.604950
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108835
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079526.1
NP_001072994.1
9
1560
NM_016260.2
NP_057344.2
8
1491
XM_005246384.4
XP_005246441.1
8
1601
XM_005246385.3
XP_005246442.1
8
1631
XM_005246386.4
XP_005246443.1
8
1482
XM_011510802.2
XP_011509104.1
9
1673
XM_011510803.2
XP_011509105.1
9
1522
XM_011510804.2
XP_011509106.1
9
1445
XM_011510805.2
XP_011509107.1
9
1534
XM_011510807.2
XP_011509109.1
9
1709
XM_011510808.2
XP_011509110.1
9
1559
XM_011510809.2
XP_011509111.1
9
1571
XM_011510810.2
XP_011509112.1
9
1485
XM_011510811.2
XP_011509113.1
8
1606
XM_011510812.2
XP_011509114.1
8
1467
XM_011510814.2
XP_011509116.1
8
1965
XM_011510815.2
XP_011509117.1
8
1413
XM_011510816.2
XP_011509118.1
6
1414
XM_011510817.2
XP_011509119.1
7
1349
XM_011510818.2
XP_011509120.1
6
7068
XM_011510819.2
XP_011509121.1
7
1338
XM_017003589.1
XP_016859078.1
8
1553
XM_017003590.1
XP_016859079.1
8
1523
XM_017003591.1
XP_016859080.1
6
1956
XM_017003592.1
XP_016859081.1
7
1270
GenBank mRNA
AF130863.1
-
8
1491
AK301503.1
-
8
1487
AY008294.1
-
2
1955
AY587062.1
-
9
1475
AY587063.1
-
9
1437
AY587064.1
-
9
1521
AY587065.1
-
8
1328
AY587066.1
-
6
1100
AY587067.1
-
7
1139
AY587068.1
-
8
1399
AY587069.1
-
7
1244
AY587070.1
-
5
887
AY587071.1
-
6
1106
AY587072.1
-
3
434
AY587073.1
-
8
1501
BC028936.1
-
9
1560
4540576
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
biological_process
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System