Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PER3 SIRNA Assays ›
Gene Symbol:
PER3
Gene Name:
period circadian clock 3
Gene Aliases:
FASPS3, GIG13
Chromosome Location:
Chr.1: 7784285 - 7845181 on Build GRCh38
UniGene ID:
Hs.162200
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108922
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PER3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289861.1
NP_001276790.1
13
1658
NM_001289862.1
NP_001276791.1
13
1658
NM_001289863.1
NP_001276792.1
13
1658
NM_001289864.1
NP_001276793.1
13
1719
NM_016831.2
NP_058515.1
13
1655
XM_005263524.3
XP_005263581.1
12
1533
XM_011542385.2
XP_011540687.1
12
1533
XM_011542390.2
XP_011540692.1
12
1533
XM_017002723.1
XP_016858212.1
12
1530
XM_017002724.1
XP_016858213.1
12
1530
XM_017002725.1
XP_016858214.1
12
1533
XM_017002726.1
XP_016858215.1
12
1530
XM_017002727.1
XP_016858216.1
14
1852
XM_017002728.1
XP_016858217.1
12
1594
XM_017002729.1
XP_016858218.1
10
1034
XM_017002730.1
XP_016858219.1
12
1594
XM_017002731.1
XP_016858220.1
12
1594
XM_017002732.1
XP_016858221.1
11
1643
XM_017002733.1
XP_016858222.1
14
1759
XM_017002734.1
XP_016858223.1
15
1901
XM_017002735.1
XP_016858224.1
14
1759
XM_017002736.1
XP_016858225.1
15
1901
XM_017002737.1
XP_016858226.1
9
1038
XM_017002738.1
XP_016858227.1
9
1044
GenBank mRNA
AB047686.1
-
12
1618
AK307760.1
-
12
1533
AK308303.1
-
13
1707
AL157954.1
-
12
1566
AY493418.1
-
12
1462
BC144137.1
-
12
1623
BC146781.1
-
12
1618
9007235
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
circadian regulation of gene expression
regulation of circadian sleep/wake cycle, sleep
protein stabilization
protein binding
kinase binding
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0