Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ENO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001976.4 | NP_001967.3 | 4 | 338 | ||
NM_053013.3 | NP_443739.3 | 4 | 296 | |||
XM_005256521.2 | XP_005256578.1 | 4 | 724 | |||
XM_011523729.1 | XP_011522031.1 | 4 | 314 | |||
XM_017024346.1 | XP_016879835.1 | 4 | 731 | |||
GenBank mRNA | AK300662.1 | - | 4 | 266 | ||
AK300673.1 | - | 4 | 308 | |||
AK309864.1 | - | 5 | 389 | |||
AU143784.1 | - | 4 | 335 | |||
BC017249.2 | - | 4 | 253 | 4902149 | ||
BJ995304.1 | - | 4 | 312 | |||
DA082822.1 | - | 4 | 296 | |||
DA566866.1 | - | 4 | 294 | |||
X16504.1 | - | 3 | 200 | |||
X51957.1 | - | 3 | 212 |