Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145343.1 | NP_001138815.1 | 5 | 2207 | ||
NM_001145344.1 | NP_001138816.1 | 5 | 2204 | |||
NM_001145345.1 | NP_001138817.1 | 5 | 2216 | |||
NM_001300970.1 | NP_001287899.1 | 4 | 2430 | |||
NM_032838.4 | NP_116227.1 | 5 | 2151 | |||
XM_005259354.2 | XP_005259411.1 | 4 | 2449 | |||
XM_005259356.2 | XP_005259413.1 | 3 | 2334 | |||
XM_006723447.2 | XP_006723510.1 | 5 | 2560 | |||
XM_011527428.1 | XP_011525730.1 | 3 | 2322 | |||
XM_017027399.1 | XP_016882888.1 | 4 | 2418 | |||
XM_017027400.1 | XP_016882889.1 | 2 | 1979 | |||
GenBank mRNA | AK027685.1 | - | 5 | 2150 | ||
AK074497.1 | - | 5 | 2162 | |||
BX649070.1 | - | 3 | 2282 |