Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC36A1 SIRNA Assays ›
Gene Symbol:
SLC36A1
Gene Name:
solute carrier family 36 member 1
Gene Aliases:
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location:
Chr.5: 151411344 - 151556318 on Build GRCh38
UniGene ID:
Hs.269004
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109926
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC36A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308150.1
NP_001295079.1
9
1080
NM_078483.3
NP_510968.2
9
1080
XM_005268386.1
XP_005268443.1
9
1050
XM_006714759.3
XP_006714822.1
9
1129
XM_011537580.2
XP_011535882.1
9
1770
XM_011537581.1
XP_011535883.1
10
1198
XM_011537583.2
XP_011535885.1
10
5635
XM_011537584.2
XP_011535886.1
9
1289
XM_011537585.1
XP_011535887.1
10
1145
XM_011537586.2
XP_011535888.1
9
1893
XM_011537587.2
XP_011535889.1
9
1149
XM_011537588.2
XP_011535890.1
9
993
XM_011537589.2
XP_011535891.1
9
1901
XM_011537590.1
XP_011535892.1
9
908
XM_011537591.1
XP_011535893.1
9
883
XM_011537592.2
XP_011535894.1
9
887
XM_011537593.2
XP_011535895.1
10
1143
XM_011537594.1
XP_011535896.1
11
1257
XM_011537595.2
XP_011535897.2
9
1776
XM_017009216.1
XP_016864705.1
9
976
XM_017009217.1
XP_016864706.1
9
1080
XM_017009218.1
XP_016864707.1
8
862
XM_017009219.1
XP_016864708.1
9
1130
GenBank mRNA
AF516142.1
-
8
872
AK057340.1
-
9
1050
AK309323.1
-
9
1052
AY162213.1
-
9
985
AY227111.1
-
10
1000
AY227112.1
-
3
368
BC136437.1
-
9
951
9020426
BC136438.1
-
9
951
9020427
BX537963.1
-
9
1077
GU296670.1
-
8
857
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System