Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLHC1 SIRNA Assays ›
Gene Symbol:
CLHC1
Gene Name:
clathrin heavy chain linker domain containing 1
Gene Aliases:
C2orf63
Chromosome Location:
Chr.2: 55172547 - 55232563 on Build GRCh38
UniGene ID:
Hs.347014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110151
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135598.1
NP_001129070.1
3
274
NM_152385.2
NP_689598.2
5
725
XM_005264136.2
XP_005264193.1
4
593
XM_006711937.3
XP_006712000.2
2
32
XM_011532519.2
XP_011530821.1
5
3618
XM_011532520.2
XP_011530822.1
5
896
XM_011532521.2
XP_011530823.1
6
1052
XM_011532522.2
XP_011530824.1
6
1039
XM_011532523.2
XP_011530825.1
5
1128
XM_011532524.1
XP_011530826.1
4
510
XM_011532525.2
XP_011530827.1
4
576
XM_011532527.2
XP_011530829.1
5
4220
XM_011532528.1
XP_011530830.1
4
1120
XM_011532529.2
XP_011530831.1
3
1079
XM_011532530.1
XP_011530832.1
3
840
XM_017003333.1
XP_016858822.1
5
3618
XM_017003334.1
XP_016858823.1
5
896
XM_017003335.1
XP_016858824.1
3
3788
XM_017003336.1
XP_016858825.1
4
1209
XM_017003337.1
XP_016858826.1
2
298
XM_017003338.1
XP_016858827.1
2
276
XM_017003339.1
XP_016858828.1
5
3618
XM_017003343.1
XP_016858832.1
4
1120
XM_017003344.1
XP_016858833.1
4
1208
XM_017003345.1
XP_016858834.1
3
1078
XM_017003346.1
XP_016858835.1
5
3618
XM_017003348.1
XP_016858837.1
5
3618
XM_017003349.1
XP_016858838.1
1
3252
XM_017003350.1
XP_016858839.1
3
1078
GenBank mRNA
AK315683.1
-
5
683
BC029502.1
-
5
725
5270720
BC036908.1
-
3
274
5166064
DA713663.1
-
4
487
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0