Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HKR1 SIRNA Assays ›
Gene Symbol:
HKR1
Gene Name:
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases:
ZNF875
Chromosome Location:
Chr.19: 37317900 - 37364455 on Build GRCh38
UniGene ID:
Hs.657027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110482
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HKR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181786.2
NP_861451.1
6
2441
XM_017026672.1
XP_016882161.1
4
2629
XM_017026673.1
XP_016882162.1
4
2695
XM_017026674.1
XP_016882163.1
8
2995
XM_017026675.1
XP_016882164.1
8
2992
XM_017026676.1
XP_016882165.1
5
2290
XM_017026677.1
XP_016882166.1
5
2277
XM_017026678.1
XP_016882167.1
5
2287
XM_017026679.1
XP_016882168.1
5
2274
XM_017026680.1
XP_016882169.1
8
2657
XM_017026681.1
XP_016882170.1
7
2605
XM_017026682.1
XP_016882171.1
8
2654
XM_017026683.1
XP_016882172.1
7
2516
XM_017026684.1
XP_016882173.1
6
2378
XM_017026685.1
XP_016882174.1
4
2223
XM_017026686.1
XP_016882175.1
7
2513
XM_017026687.1
XP_016882176.1
6
2375
XM_017026688.1
XP_016882177.1
4
4447
XM_017026689.1
XP_016882178.1
6
2461
XM_017026690.1
XP_016882179.1
4
2241
XM_017026691.1
XP_016882180.1
3
2021
XM_017026692.1
XP_016882181.1
8
2591
XM_017026693.1
XP_016882182.1
9
2712
XM_017026694.1
XP_016882183.1
9
2662
XM_017026695.1
XP_016882184.1
8
2634
XM_017026696.1
XP_016882185.1
7
2486
XM_017026697.1
XP_016882186.1
6
2408
XM_017026698.1
XP_016882187.1
6
2386
XM_017026699.1
XP_016882188.1
4
5222
XM_017026700.1
XP_016882189.1
6
2387
XM_017026701.1
XP_016882190.1
8
2585
XM_017026702.1
XP_016882191.1
7
2507
XM_017026703.1
XP_016882192.1
5
2284
XM_017026704.1
XP_016882193.1
5
2346
XM_017026705.1
XP_016882194.1
2
2343
XM_017026707.1
XP_016882196.1
5
2759
XM_017026708.1
XP_016882197.1
5
2762
GenBank mRNA
AB013897.1
-
5
2288
AK056243.1
-
6
2375
AK128116.1
-
9
2994
AK223616.1
-
5
2280
AK301601.1
-
5
1736
BC004513.2
-
1
1463
BC017256.2
-
6
2441
3356680
BC053845.1
-
4
2278
5493685
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-h42lq:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0