Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-DQB1 SIRNA Assays ›
          Gene Symbol:
          HLA-DQB1
          Gene Name:
          major histocompatibility complex, class II, DQ beta 1
          Gene Aliases:
          CELIAC1, HLA-DQB, HLA-DRB1, IDDM1
          Chromosome Location:
          Chr.6: 32659464 - 32666689 on Build GRCh38
          UniGene ID:
          Hs.409934
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 11053

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-DQB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243962.1 NP_001230891.1 1, 2 198
          GenBank mRNA AB209638.1 - 1, 2 178
          AF113250.1 - 1 17
          AF195245.1 - 1 23
          AF291674.1 - 1, 2 119
          AK295255.1 - 1, 2 166
          AK295476.1 - 1, 2 171
          AK295695.1 - 1, 2 169
          AK296060.1 - 1, 2 163
          AK296613.1 - 1, 2 180
          AK304543.1 - 1, 2 205
          AY656681.1 - 1, 2 119
          AY656682.1 - 1, 2 119
          BC012106.1 - 1, 2 173 4574913
          DQ136036.1 - 1, 2 164
          J00198.1 - 1 28
          KC879155.1 - 11
          L34095.1 - 1 23
          L34096.1 - 1 23
          L34097.1 - 1 23
          L34098.1 - 1 23
          L34101.1 - 1 23
          L34102.1 - 1 23
          L34103.1 - 1 23
          L34106.1 - 1 23
          L34107.1 - 1 23
          L40179.1 - 1 23
          L40180.1 - 1 23
          L42626.1 - 1 23
          M14189.1 - 1 23
          M16996.1 - 1, 2 119
          M17206.1 - 1 23
          M17563.1 - 1, 2 131
          M17564.1 - 1, 2 131
          M17565.1 - 1, 2 87
          M17955.1 - 1, 2 191
          M19239.1 - 1 23
          M20505.1 - 1, 2 87
          M21135.1 - 1 23
          M24364.1 - 1, 2 163
          M25325.1 - 1, 2 201
          M25326.1 - 1, 2 161
          M32577.1 - 1, 2 87
          M35982.1 - 1, 2 102
          M59800.1 - 1 23
          M59801.1 - 1 23
          M60028.1 - 1, 2 176
          M81140.1 - 1, 2 140
          M81141.1 - 1, 2 154
          M83770.1 - 1 23
          S79787.1 - 11
          U63321.1 - 1 17
          U66400.1 - 1 23
          U83582.1 - 1, 2 117
          U83585.1 - 1, 2 117
          V00529.1 - 1 28
          X00369.1 - 1, 2 87
          X03068.1 - 1, 2 200
          X76553.1 - 1 23
          X76554.1 - 1 23
          X96420.1 - 1, 2 119

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immunoglobulin production involved in immunoglobulin mediated immune response
          humoral immune response mediated by circulating immunoglobulin
          immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          T cell receptor signaling pathway
          interferon-gamma-mediated signaling pathway
          MHC class II receptor activity
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0