Search Thermo Fisher Scientific
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000098.2 | NP_000089.1 | 3, 4 | 852 | ||
XM_005270484.1 | XP_005270541.1 | 3, 4 | 520 | |||
GenBank mRNA | AK223052.1 | - | 1, 2 | 95 | ||
AK312687.1 | - | 3, 4 | 456 | |||
M58581.1 | - | 3, 4 | 438 | |||
U09648.1 | - | 3, 4 | 853 |