Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYB5R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000398.6 | NP_000389.1 | 6 | 573 | ||
NM_001129819.2 | NP_001123291.1 | 6 | 817 | |||
NM_001171660.1 | NP_001165131.1 | 6 | 713 | |||
NM_001171661.1 | NP_001165132.1 | 7 | 633 | |||
NM_007326.4 | NP_015565.1 | 6 | 572 | |||
GenBank mRNA | AF361370.1 | - | 6 | 508 | ||
AJ010116.1 | - | 6 | 486 | |||
AJ010117.1 | - | 6 | 486 | |||
AJ010118.1 | - | 6 | 486 | |||
AJ310899.1 | - | 3 | 190 | |||
AJ310900.1 | - | 3 | 190 | |||
AK124528.1 | - | 3 | 2623 | |||
AK302204.1 | - | 6 | 712 | |||
AY421733.1 | - | 1 | 36 | |||
BC004821.1 | - | 6 | 517 | 3450773 | ||
BM564146.1 | - | 7 | 623 | |||
BT009821.1 | - | 6 | 489 | |||
BX334220.2 | - | 6 | 574 | |||
BX366240.2 | - | 6 | 573 | |||
CB118906.1 | - | 6 | 561 | |||
CR456435.1 | - | 6 | 495 | |||
DA268749.1 | - | 6 | 507 | |||
DA966919.1 | - | 6 | 507 | |||
DB029897.1 | - | 6 | 572 | |||
M16461.1 | - | 5 | 470 | |||
M16462.1 | - | 5 | 408 | |||
Y09501.1 | - | 6 | 508 |