Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11091
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
2
492
NM_001256765.1
NP_001243694.1
3
441
NM_002189.3
NP_002180.1
2
288
NM_172200.2
NP_751950.2
2
288
XM_011519461.2
XP_011517763.1
4
682
XM_011519462.2
XP_011517764.1
3
600
XM_011519463.2
XP_011517765.1
4
682
XM_011519464.2
XP_011517766.1
3
529
XM_011519465.1
XP_011517767.1
4
682
XM_011519466.2
XP_011517768.1
4
682
XM_011519467.2
XP_011517769.1
2
445
XM_011519468.1
XP_011517770.1
4
682
XM_011519469.1
XP_011517771.1
4
682
XM_011519470.2
XP_011517772.1
2
447
XM_017016195.1
XP_016871684.1
3
529
XM_017016196.1
XP_016871685.1
2
254
XM_017016197.1
XP_016871686.1
3
269
XM_017016199.1
XP_016871688.1
4
682
XM_017016200.1
XP_016871689.1
2
939
XM_017016201.1
XP_016871690.1
3
636
XM_017016202.1
XP_016871691.1
3
258
XM_017016203.1
XP_016871692.1
2
939
GenBank mRNA
AF035279.1
-
2
223
AK304211.1
-
2
199
BC074726.2
-
2
200
30915179
BC107777.1
-
2
356
6497994
BC121140.2
-
2
195
40120998
BC121141.2
-
2
200
40120999
CR457064.1
-
1
61
CR542023.1
-
2
169
DC298692.1
-
2
293
HQ401283.1
-
3
322
JX987304.1
-
3
322
JX987305.1
-
3
322
JX987306.1
-
3
322
JX987307.1
-
3
322
JX987308.1
-
3
322
JX987309.1
-
3
322
JX987310.1
-
3
322
JX987311.1
-
3
322
U31628.1
-
2
251
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0