Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGA2 SIRNA Assays ›
          Gene Symbol:
          ITGA2
          Gene Name:
          integrin subunit alpha 2
          Gene Aliases:
          BR, CD49B, GPIa, HPA-5, VLA-2, VLAA2
          Chromosome Location:
          Chr.5: 52989326 - 53094779 on Build GRCh38
          UniGene ID:
          Hs.482077
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111111

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITGA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002203.3 NP_002194.2 5 562
          NR_073103.1 5 562
          NR_073104.1 5 562
          NR_073105.1 5 562
          NR_073106.1 5 562
          NR_073107.1 4 441
          GenBank mRNA AK307952.1 - 5 551
          BC143505.1 - 5 448
          BC143508.1 - 5 448
          BC143509.1 - 5 448
          BC143511.1 - 5 448
          BC143512.1 - 4 327
          BP280760.1 - 5 565
          X17033.1 - 5 467

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of leukocyte migration
          substrate-dependent cell migration
          hypotonic response
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          female pregnancy
          blood coagulation
          cell proliferation
          organ morphogenesis
          positive regulation of epithelial cell migration
          positive regulation of alkaline phosphatase activity
          response to amine
          response to muscle activity
          positive regulation of smooth muscle cell migration
          extracellular matrix organization
          mammary gland development
          positive regulation of cell projection organization
          cell-substrate adhesion
          positive regulation of collagen biosynthetic process
          positive regulation of collagen binding
          response to L-ascorbic acid
          cell adhesion mediated by integrin
          collagen-activated signaling pathway
          response to drug
          positive regulation of DNA binding
          skin morphogenesis
          establishment of protein localization
          positive regulation of translation
          positive regulation of cell adhesion
          positive regulation of smooth muscle contraction
          viral entry into host cell
          focal adhesion assembly
          mesodermal cell differentiation
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of positive chemotaxis
          detection of mechanical stimulus involved in sensory perception of pain
          positive regulation of transmission of nerve impulse
          positive regulation of phagocytosis, engulfment
          hepatocyte differentiation
          response to parathyroid hormone
          cellular response to mechanical stimulus
          cellular response to estradiol stimulus
          virus receptor activity
          integrin binding
          protein binding
          collagen binding
          protein complex binding
          collagen receptor activity
          laminin binding
          metal ion binding
          protein heterodimerization activity
          collagen binding involved in cell-matrix adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0