Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST6GAL1 SIRNA Assays ›
          Gene Symbol:
          ST6GAL1
          Gene Name:
          ST6 beta-galactoside alpha-2,6-sialyltransferase 1
          Gene Aliases:
          SIAT1, ST6GalI, ST6N
          Chromosome Location:
          Chr.3: 186930343 - 187078553 on Build GRCh38
          UniGene ID:
          Hs.207459
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111233

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST6GAL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003032.2 NP_003023.1 5 1092
          NM_173216.2 NP_775323.1 6 1415
          NM_173217.2 NP_775324.1 5 758
          XM_005247719.1 XP_005247776.1 5 1003
          XM_005247720.1 XP_005247777.1 5 1061
          XM_006713734.1 XP_006713797.1 7 1670
          XM_011513085.1 XP_011511387.1 7 1678
          XM_011513086.1 XP_011511388.1 5 1061
          XM_017007065.1 XP_016862554.1 8 1762
          XM_017007066.1 XP_016862555.1 5 1090
          XM_017007067.1 XP_016862556.1 5 1210
          XM_017007068.1 XP_016862557.1 5 1095
          GenBank mRNA A17362.1 - 6 1219
          A23699.1 - 3 796
          AK128726.1 - 1 952
          AK292879.1 - 5 1113
          AK312023.1 - 2 121
          BC031476.1 - 5 1086 4339124
          BC040009.1 - 5 1092 5557868
          DB235666.1 - 5 547
          X17247.1 - 6 1213
          X54363.1 - 5 1051
          X62822.1 - 5 1092

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          N-acetylneuraminate metabolic process
          humoral immune response
          oligosaccharide metabolic process
          O-glycan processing
          protein N-linked glycosylation via asparagine
          positive regulation of mononuclear cell proliferation
          response to ethanol
          negative regulation of chemotaxis
          sialylation
          regulation of substrate adhesion-dependent cell spreading
          protein sialylation
          negative regulation of macrophage apoptotic process
          beta-galactoside alpha-2,6-sialyltransferase activity
          protein binding
          sialyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0