Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPFIA1 SIRNA Assays ›
          Gene Symbol:
          PPFIA1
          Gene Name:
          PTPRF interacting protein alpha 1
          Gene Aliases:
          LIP.1, LIP1, LIPRIN
          Chromosome Location:
          Chr.11: 70270687 - 70384501 on Build GRCh38
          UniGene ID:
          Hs.530749
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111327

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPFIA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003626.3 NP_003617.1 3 497
          NM_177423.2 NP_803172.1 3 497
          NR_045286.1 3 497
          XM_006718716.3 XP_006718779.1 3 510
          XM_011545306.2 XP_011543608.1 3 382
          XM_011545307.2 XP_011543609.1 3 510
          XM_011545308.2 XP_011543610.1 3 510
          XM_011545309.2 XP_011543611.1 3 510
          XM_011545310.2 XP_011543612.1 3 510
          XM_011545311.2 XP_011543613.1 3 510
          XM_011545312.2 XP_011543614.1 3 510
          XM_011545313.2 XP_011543615.1 3 510
          XM_011545314.2 XP_011543616.1 3 510
          XM_011545315.2 XP_011543617.1 3 510
          XM_011545316.2 XP_011543618.1 3 510
          XM_011545317.2 XP_011543619.1 3 510
          XM_011545319.1 XP_011543621.1 3 510
          XM_017018448.1 XP_016873937.1 3 510
          XM_017018449.1 XP_016873938.1 3 510
          XM_017018450.1 XP_016873939.1 3 510
          GenBank mRNA AB209314.1 - 3 424
          AW163189.1 - 3 497
          BC034046.1 - 3 460 4794300
          U22815.1 - 3 511
          U22816.1 - 3 511

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          signal transduction
          neurotransmitter secretion
          glutamate secretion
          negative regulation of stress fiber assembly
          negative regulation of establishment of protein localization to plasma membrane
          signal transducer activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline