Search
Search

SHMT1| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281786.1 | NP_001268715.1 | 11 | 1797 | ||
| NM_004169.4 | NP_004160.3 | 12 | 1913 | |||
| NM_148918.2 | NP_683718.1 | 11 | 1796 | |||
| XM_005256767.3 | XP_005256824.1 | 13 | 1984 | |||
| XM_011523992.2 | XP_011522294.1 | 10 | 1641 | |||
| XM_017024957.1 | XP_016880446.1 | 12 | 1738 | |||
| XM_017024958.1 | XP_016880447.1 | 12 | 1867 | |||
| XM_017024959.1 | XP_016880448.1 | 9 | 1488 | |||
| XM_017024960.1 | XP_016880449.1 | 8 | 1365 | |||
| GenBank mRNA | AK223552.1 | - | 12 | 1844 | ||
| AK298415.1 | - | 11 | 1729 | |||
| BC007979.2 | - | 12 | 1759 | 3687873 | ||
| BC038598.1 | - | 12 | 1770 | 4523709 | ||
| BM792235.1 | - | 1 | 40 | |||
| Y14486.1 | - | 7 | 2025 | |||