Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRDMT1 SIRNA Assays ›
          Gene Symbol:
          TRDMT1
          Gene Name:
          tRNA aspartic acid methyltransferase 1
          Gene Aliases:
          DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
          Chromosome Location:
          Chr.10: 17136785 - 17202071 on Build GRCh38
          UniGene ID:
          Hs.351665
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111448

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRDMT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321006.1 NP_001307935.1 2 513
          NM_001321007.1 NP_001307936.1 2 513
          NM_004412.6 NP_004403.1 2 513
          XM_005252372.4 XP_005252429.1 2 508
          XM_005252374.4 XP_005252431.1 2 508
          XM_005252375.4 XP_005252432.1 2 508
          XM_005252378.3 XP_005252435.1 2 109
          XM_006717388.3 XP_006717451.1 2 508
          XM_011519343.2 XP_011517645.1 2 508
          XM_011519344.2 XP_011517646.1 2 508
          XM_011519345.2 XP_011517647.1 2 921
          XM_017015800.1 XP_016871289.1 2 508
          XM_017015801.1 XP_016871290.1 2 508
          XM_017015802.1 XP_016871291.1 1 12
          XM_017015803.1 XP_016871292.1 2 105
          XM_017015804.1 XP_016871293.1 2 112
          XM_017015806.1 XP_016871295.1 2 109
          XM_017015807.1 XP_016871296.1 2 130
          XM_017015808.1 XP_016871297.1 2 107
          XM_017015809.1 XP_016871298.1 2 508
          XM_017015810.1 XP_016871299.1 2 109
          GenBank mRNA AF012128.1 - 2 83
          AF045888.1 - 2 82
          AF329940.1 - 1 22
          AF329941.1 - 1 22
          AF329942.1 - 1 22
          AF329943.1 - 1 22
          AF329944.1 - 1 22
          AJ223333.1 - 2 124
          AK291417.1 - 2 82
          AK299027.1 - 2 293
          AK303428.1 - 2 99
          BC047733.1 - 2 83 6473993
          BX537961.1 - 2 103
          CR450316.1 - 2 76
          CV816165.1 - 2 496
          DC411638.1 - 2 513
          HQ637546.1 - 2 76
          HQ637547.1 - 2 76
          HQ637548.1 - 2 76
          HQ637549.1 - 2 76

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          tRNA modification
          tRNA methylation
          C-5 methylation of cytosine
          RNA binding
          DNA (cytosine-5-)-methyltransferase activity
          tRNA methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0